Learn More
Waldmann Lighting™ Sealed Flourescent Tube Light
Provides broad, wide area illumination inside the machine compartment. The Waldmann Lighting™ RL70CE-124 and RL70CE-136 fluorescent tube lights have a strong acrylic tube and energy-efficient compact fluorescent lamp. 2.75 in. diameter.
$465.16 - $487.20
Specifications
Includes Lamp
Light Source CFL
Certifications/Compliance ETL/cETL, CE
Voltage 120/277 V
Material Acrylic Tube
View More Specs
Products 2
Catalog Number Mfr. No. Wattage Length (English) Lumen Price Quantity & Availability  
Catalog Number Mfr. No. Wattage Length (English) Lumen Price Quantity & Availability  
11-000-125
Encompass
View Documents
Waldmann Lighting™
10867000000081448
24 W 26.7 in. 2000 lm
Each for $487.20
Add to cart
11-000-126
Encompass
View Documents
Waldmann Lighting™
10868000000081455
36 W 31 in. 2900 lm
Each for $465.16
Add to cart
Description
Description
  • Designed to be impervious to debris, coolants and lubricants
  • Ready to mount into existing configurations
  • Offers up to 10,000 lumens of super bright light output
  • Brackets are necessary to mount these lights
  • Offers up to 10,000 lumens of super bright light output 4100K color temperature
  • Electronic ballasts offer flicker-free, energy-efficient performance
  • 2.75-inch outside diameter tubes
Specifications
Specifications
Lamp
ETL/cETL, CE
Acrylic Tube
CFL
120/277 V
2.75 in.