Learn More
BrandTech™ Sealing films for microplates and PCR plates
Premium sealing films for microplates, deepwell plates and PCR plates
Manufacturer:  BrandTech™ 701390ES
 View more versions of this product
Catalog No. 14-380-920
Add to cart
Description
Description
  • Polyester film is optimized for ELISA applications, and is available plain, with colored strips for plate tracking, or in strips for easy well access
  • Vinyl film that is piereceable/resealable for use in automation applications.
  • Polyolefin film is highly transparent and suitable for real time qPCR and protein crystallization. It features a special non-tacky surface for easy placement and reduced chance of sample contaimination.
Specifications
Specifications
deep well plates
ANSI/SLAS
clear
Pack of 100 sheets
sealing film
polyester
No