Learn More
Innovating Science™ Seamless Cellulose Dialysis Tubing Products for Science Education
Retains materials with 12,000 and higher molecular weight.
Products 1
Catalog Number Mfr. No. Description Price Qty Availability  
Catalog Number Mfr. No. Description Price Qty Availability  
S413002 Products for Science Education
View Documents
Innovating Science™
DD010610FT
N/A 15.9mm inflated dia., 25mm flat; 10 ft. L roll (3m) This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
  • Especially useful in analytical chemistry, biology, and pharmaceutical fields
  • Highly permeable to water and low molecular weight compounds
  • Contains glycerin, water, 1% sulfur
  • Must be stored in air-tight container or rehumidified before use
  • Pore diameter: 4.8nm
† Average values.