Learn More
Laxco™ SeBa™ 2 Series Digital Microscope System Products for Science Education
Laxco™ SeBa™ 2 Series Digital Microscope System integrates digital technology with high quality microscope optics into seamless easy-to-use system.
$4216.00 - $5328.00
Specifications
Magnification Power 4X, 10X, 40X, 100X oil
Optical Design Infinity, DIN
Condenser Type N.A. 1.25 Abbe Achromatic condenser with Iris diaphragm and filter holder
Lighting Type 3w LED
Product Type Specs Microscope
View More Specs
Products 2
Catalog Number Mfr. No. Model Design Type Price Quantity & Availability  
Catalog Number Mfr. No. Model Design Type Price Quantity & Availability  
S72274 Products for Science Education
View Documents
Laxco™
SEBA2BF1/EMD
SeBa2-BF1 Contrast Method: Brightfield
Each for $4,216.00
Add to cart
S72272 Products for Science Education
View Documents
Laxco™
SEBA2PH1/EMD
SeBa2-PH1 Contrast Method: Phase Contrast
Each for $5,328.00
Add to cart
Description
Description
Head
  • 9.7 in. touchscreen retina display
  • 5MP color CMOS camera
  • 4 USB ports for attaching keyboard, mouse, storage device or remote capture button
  • SeBaAccess technology allows for individual log in, via an access card swipped in front of the Seba label
  • SeBaShare cloud based storage allows simple uploading of images to unique account on the cloud for accessing of images anywhere (log in based off from SeBaAccess)
  • Capture still or video images and save them to SeBaShare or USB storage device
  • Capture, review, measure, and annotate images
  • Single power source controls power to the camera, computer and microscope
Objectives
  • Sealed 40X to prevent accidental damage from use of oil at 100X
  • Infinity corrected
  • Parfocal and parcentered
Stand
  • Built-in carrying handle
  • Reverse objective turret (4 position) allows for easy access to slides
  • Adjustable focusing stop protects slides from accidental damage
  • Removable LED cartridge
Illumination
  • Full Koehler illumination with field diaphragm
  • Cool, stable, color temperature and brightness intensity
  • 50,000 bulb life
Specifications
Specifications
4X, 10X, 40X, 100X oil
N.A. 1.25 Abbe Achromatic condenser with Iris diaphragm and filter holder
Microscope
150x132mm X/Y Mechanical Stage, Moving Range: 76x50mm
4 USB Ports, Bluetooth, Wifi
Reversed Quadruple Nosepiece
Infinity, DIN
3w LED
Coarse and Fine
5MPcmOS, 2.2andmu;m pixels
Capture still images and video, Upload to SeBaShare
9.7 in. HD Touchscreen Display 1024 x 768 Pixels