Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Laxco™ SeBa™ 3 Microscope, Routine Clinical Configuration

Laxco™ SeBa™ 3 Series Digital Microscope System integrates digital technology with high quality microscope optics into seamless easy-to-use system for routine clinical applications

Manufacturer:  Laxco™ SEBA3RC3

Catalog No. SEBA3RC3


Add to cart

Description

Description

  • 5 position objective nosepiece
  • 9.7 in. full retina touchscreen display
  • 5MP color camera
  • Individual user login via access card(s)
  • Easily capture, review, measure, print, and annotate images
  • Brightfield, darkfield, simple polarization, and phase contrast
  • Upload images to SeBaShare or save to USB drive
  • User-friendly touchscreen interface
  • Parfocal, parcentric, infinity plan achro optical system
Specifications

Specifications

SEBA 9.7 in. Display
Brightfield
4X, 10X, 40X, 100X Oil Plan Achromat
3w LED
1.25 N.A. ABBE
Infinity, 22mm FOV
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit