Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sebacic Acid, 98%, ACROS Organics™

Click to view available options
Quantity:
100g
1kg
2.5kg
10kg
Packaging:
Plastic bottle
Plastic drum
111-20-6
C10H18O4
202.25
MFCD00004440
CXMXRPHRNRROMY-UHFFFAOYSA-N
sebacic acid,1,8-octanedicarboxylic acid,1,10-decanedioic acid,sebacic acids,decanedicarboxylic acid,n-decanedioic acid,sebacinsaure,acide sebacique,sebacate,sebacinsaeure
5192
CHEBI:41865
decanedioic acid
C(CCCCC(=O)O)CCCC(=O)O
111-20-6
C10H18O4
202.25
MFCD00004440
CXMXRPHRNRROMY-UHFFFAOYSA-N
sebacic acid,1,8-octanedicarboxylic acid,1,10-decanedioic acid,sebacic acids,decanedicarboxylic acid,n-decanedioic acid,sebacinsaure,acide sebacique,sebacate,sebacinsaeure
5192
CHEBI:41865
decanedioic acid
C(CCCCC(=O)O)CCCC(=O)O
Color White
Water 0.3% max.
Packaging Plastic bottle
Flash Point 220°C
Melting Point 131.0°C to 134.0°C
Boiling Point 294.4°C (100.0 mmHg)
Infrared Spectrum Authentic
Quantity 2.5kg
Assay Percent Range 98%
Linear Formula HO2C(CH2)8CO2H
Beilstein 02, IV, 2078
Merck Index 15, 8552
Solubility Information Solubility in water: 1g/L (20°C). Other solubilities: 0.1% in ether (17°C), freely soluble in alcohol, esters and ketones, sparingly soluble in hydrocarbons, insoluble in ethyl ether and carbon tetrachloride
Formula Weight 202.25
Physical Form Powder or Granules
Percent Purity 98%
Chemical Name or Material Sebacic acid
Show More Show Less