Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Sebaconitrile, 95%

Click to view available options
Quantity:
5g
25g
100g
1871-96-1
C10H16N2
164.252
MFCD00019900
DFJYZCUIKPGCSG-UHFFFAOYSA-N
sebaconitrile,1,8-dicyanooctane,octamethylene dicyanide,oktamethylendikyanid,oktamethylendikyanid czech,octane-1,8-dicarbonitrile,sebacic acid dinitrile,acmc-209epg,wln: nc8cn,4-02-00-02089 beilstein handbook reference
74639
decanedinitrile
C(CCCCC#N)CCCC#N
1871-96-1
C10H16N2
164.252
MFCD00019900
DFJYZCUIKPGCSG-UHFFFAOYSA-N
sebaconitrile,1,8-dicyanooctane,octamethylene dicyanide,oktamethylendikyanid,oktamethylendikyanid czech,octane-1,8-dicarbonitrile,sebacic acid dinitrile,acmc-209epg,wln: nc8cn,4-02-00-02089 beilstein handbook reference
74639
decanedinitrile
C(CCCCC#N)CCCC#N
Quantity 100g
Flash Point >110°C (230°F)
Melting Point 4°C to 8°C
Boiling Point 199°C to 200°C
Beilstein 1750739
Formula Weight 164.25
Percent Purity 95%
Chemical Name or Material Sebaconitrile

GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P280-P301+P312-P312-P363-P322-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Specific measures (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : VS1500000

TSCA : TSCA