Alfa Aesar™ sec-Butyl methyl sulfide, 98%

Manufacturer: Alfa Aesar™ B2194214

Quantity 25g

 View more versions of this product

Catalog No. AAB2194214

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10359-64-5
    Molecular Formula C5H12S
    Formula Weight 104.22
    MDL Number MFCD00015193
    UN Number UN1993
    Beilstein 1730946
    Synonym Methyl sec-butyl sulfide; 2-(Methylthio)butane
    Chemical Name or Material sec-Butyl methyl sulfide

    Specifications

    Formula Weight 104.22
    Health Hazard 3 GHS P Statement
    P210-P261-P243-P403
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Take precautionary measures against static discharge.
    Store in a well-ventilated place.
    MDL Number MFCD00015193
    UN Number UN1993
    Health Hazard 1 Danger
    Boiling Point 116° to 117°C
    Flash Point 5°C (41°F)
    Molecular Formula C5H12S
    CAS 10359-64-5
    Beilstein 1730946
    Synonym Methyl sec-butyl sulfide; 2-(Methylthio)butane
    Health Hazard 2 GHS H Statement
    H225
    Highly flammable liquid and vapour.
    Quantity 25g
    Odor Unpleasant
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    Chemical Name or Material sec-Butyl methyl sulfide
    Assay 98%

    GHS H Statement
    H225
    Highly flammable liquid and vapour.

    GHS P Statement
    P210-P261-P243-P403
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Take precautionary measures against static discharge.
    Store in a well-ventilated place.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit