Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

sec-Butyllithium, 1.3M in cyclohexane, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific™

Catalog Number AA43028AE
Click to view available options
Quantity:
100 mL
500 mL
598-30-1
C4H9Li
64.06
MFCD00009323
VATDYQWILMGLEW-UHFFFAOYNA-N
sec-butyllithium, s-buli, s-butyllithium, s-butyl lithium, unii-5yv3gii1tb, lithium, 1-methylpropyl, 5yv3gii1tb, secbutyllithium, b-butyllithium, sec-butyllitium
102446
(butan-2-yl)lithium
[Li]C(C)CC
598-30-1
C4H9Li
64.06
MFCD00009323
VATDYQWILMGLEW-UHFFFAOYNA-N
sec-butyllithium, s-buli, s-butyllithium, s-butyl lithium, unii-5yv3gii1tb, lithium, 1-methylpropyl, 5yv3gii1tb, secbutyllithium, b-butyllithium, sec-butyllitium
102446
(butan-2-yl)lithium
[Li]C(C)CC

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY
Color Yellow
Density 0.750g/mL
Odor Characteristic
Packaging ChemSeal™ bottles
Linear Formula CH3CH2CH(CH3)Li
Quantity 100 mL
UN Number UN3394
Beilstein 3587206
Sensitivity Air & Moisture Sensitive
Formula Weight 64.06
Physical Form Liquid
Chemical Name or Material sec-Butyllithium
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
H224-H250-H260-H252-H304-H314-H318-H336
Extremely flammable liquid and vapor.
Catches fire spontaneously if exposed to air.
In contact with water releases flammable gases which may ignite spontaneously.
Self-heating in large quantities; may catch fire.
May be fatal if swallowed and enters airways.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause drowsiness or dizziness.
Precautionary Statement GHS P Statement
P210-P222-P301+P310-P303+P361+P353-P305+P351+P338-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not allow contact with air.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 4.2; Packaging Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, PYROPHORIC, WATER-REACTIVE
TSCA TSCA
storageNote1 2° to 8°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit