Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
sec-Butyllithium, 1.3M sol. in cyclohexane/hexane (92/8), AcroSeal™, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Colorless |
Linear Formula | C2H5CH(CH3)Li |
Fieser | 09,87; 10,75; 12,97; 16,56 |
Density | 0.7500g/mL |
Solubility Information | Solubility in water: reacts (forming lioh and butane). |
Formula Weight | 64.06 |
Percent Purity | 10.5 to 12.5% (as BuLi) (Active base) |
Packaging | AcroSeal™ Glass bottle |
Specific Gravity | 0.75 |
Quantity | 100 mL |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS P Statement May cause drowsiness or dizziness. Very toxic to aquatic life with long lasting effects. Causes severe skin burns and eye damage. May be fatal if swallowed and enters airways. Suspected of damaging fertility. May cause damage to organs through prolonged or repeated exposure if inhaled. Highly flammable liquid and vapor. In contact with water releases flammable gases which may ignite spontaneously. Catches fire spontaneously if exposed to air. Reacts violently with water. |
Precautionary Statement | GHS P Statement Do not breathe dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Do not allow contact with air. |
EINECSNumber | 209-927-7 |
TSCA | TSCA |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction