Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SEC11C Rabbit anti-Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549487

Catalog No. PA549487


Add to cart

Description

Description

Component of the microsomal signal peptidase complex which removes signal peptides from nascent proteins as they are translocated into the lumen of the endoplasmic reticulum.
Specifications

Specifications

SEC11C
Polyclonal
PBS with 0.09% sodium azide
Q9D8V7
Signal peptidase complex catalytic subunit SEC11C, Microsomal signal peptidase 21 kDa subunit, SPase 21 kDa subunit, SEC11 homolog C, SEC11-like protein 3, SPC21, SEC11C, SEC11L3, SPC21, SPCS4C
Rabbit
IgG
50 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
66286
Western Blot
Unconjugated
Sec11c
Liquid
SEC11C
KLH conjugated synthetic peptide between 85-120 amino acids from the Central region of human SEC11C.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit