Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human SEC13 Antibody, R&D Systems™
Rabbit Polyclonal Antibody
$172.84 - $520.84
Specifications
Antigen | SEC13 |
---|---|
Dilution | Western Blot 0.05 ug/mL, Simple Western 0.5 ug/mL, Immunocytochemistry 1-15 ug/mL |
Applications | Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
SEC13 Polyclonal specifically detects SEC13 in Human samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.Specifications
SEC13 | |
Western Blot, Immunocytochemistry | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
D3S1231E, D3S1231ESEC13L1npp-20, Npp-20, protein SEC13 homolog, SEC13 (S. cerevisiae)-like 1, SEC13 homolog (S. cerevisiae), SEC13L1, SEC13-like 1 isoform, SEC13-like protein 1, SEC13R, SEC13-related protein, SEC13RSEC13-like 1 (S. cerevisiae) | |
E. coli-derived recombinant human SEC13 Gly123-Gln322 Accession # P55735 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human SEC13 in direct ELISA and Western blots. |
Western Blot 0.05 ug/mL, Simple Western 0.5 ug/mL, Immunocytochemistry 1-15 ug/mL | |
Polyclonal | |
Rabbit | |
Human | |
P55735 | |
6396 | |
IgG | |
Antigen Affinity-purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title