Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SEC14L2 Mouse anti-Canine, Human, Mouse, Clone: OTI4H2, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA503723

Catalog No. 50-167-4547


Add to cart

Description

Description

This gene encodes a cytosolic protein which belongs to a family of lipid-binding proteins including Sec14p, alpha-tocopherol transfer protein, and cellular retinol-binding protein. The encoded protein stimulates squalene monooxygenase which is a downstream enzyme in the cholesterol biosynthetic pathway. Alternatively spliced transcript variants encoding different isoforms have been identified for this gene.
Specifications

Specifications

SEC14L2
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
O76054, Q99J08
C22orf6, SPF, TAP, TAP1
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
23541, 477539, 67815
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI4H2
Unconjugated
SEC14L2
Liquid
SEC14L2
Full length human recombit protein of human SEC14L2 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Canine, Human, Mouse
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit