Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SEC14L4 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548541

Catalog No. PA548541

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Probable hydrophobic ligand-binding protein; may play a role in the transport of hydrophobic ligands like tocopherol, squalene and phospholipids.
Specifications

Specifications

SEC14L4
Polyclonal
PBS with 0.09% sodium azide
Q9UDX3
SEC14-like protein 4, Tocopherol-associated protein 3, SEC14L4, TAP3
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
284904
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
SEC14L4
Liquid
SEC14L4
KLH conjugated synthetic peptide between 41-70 amino acids from the N-terminal region of human SEC14L4.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit