Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SEC22B Sheep anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Sheep Polyclonal Antibody has been used in 1 publication
$153.04 - $436.00
Specifications
Antigen | SEC22B |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 10-50 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 10-50 ug/ml, Immunohistochemistry-Frozen 10-50 ug/ml |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10098766
|
Novus Biologicals
NB10098766 |
0.5 mg |
Each for $436.00
|
|
NB017948
|
Novus Biologicals
NB100987660.1MG |
0.1 mg |
Each for $153.04
|
|
Description
SEC22B Polyclonal specifically detects SEC22B in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
SEC22B | |
Western Blot 10-50 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 10-50 ug/ml, Immunohistochemistry-Frozen 10-50 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
PBS with No Preservative | |
ERS24, ERS-24sec22b, SEC22 vesicle trafficking protein homolog B (S. cerevisiae), SEC22 vesicle trafficking protein homolog B (S. cerevisiae) (gene/pseudogene), SEC22 vesicle trafficking protein-like 1, SEC22 vesicle trafficking protein-like 1 (S. cerevisiae), SEC22 vesicle-trafficking protein homolog B, SEC22 vesicle-trafficking protein-like 1, SEC22, vesicle trafficking protein (S. cerevisiae)-like 1, SEC22L1ER-Golgi SNARE of 24 kDa, vesicle-trafficking protein SEC22b | |
SEC22B | |
IgG | |
IgG purified | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
LYOPH | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Sheep | |
Human, Mouse, Rat | |
O75396 | |
9554 | |
A synthetic peptide as a part of human, rat, mouse, hamster, orangutan, chicken Vesicle-trafficking protein SEC22b conjugated to an immunogenic carrier protein was used as the immunogen. | |
Primary | |
Reconstitute with 0.5 ml sterilized water. Add glycerol (1:1) for additional stability. | |
SEC22B. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title