Learn More
Aviva Systems Biology Corp SEC22C Antibody - middle region (ARP50146_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP50146_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SEC22C. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SEC22C. Alternate names: DKFZp761F2321, MGC13261, MGC5373, SEC22L3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-60269