Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SEC23B Monoclonal Antibody (OTI8G4), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA527262
Description
SEC23B Monoclonal antibody specifically detects SEC23B in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
SEC23B | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q15437 | |
SEC23B | |
Human recombinant protein fragment corresponding to amino acids 392-641 of SEC23B produced in E.coli | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
OTI8G4 | |
Unconjugated | |
SEC23B | |
CDAII; CDA-II; CDAN2; CWS7; HEMPAS; hSec23B; Protein transport protein Sec23B; Sec23 homolog B; Sec23 homolog B (S. cerevisiae); Sec23 homolog B, coat complex II component; Sec23 homolog B, COPII coat complex component; Sec23b; SEC23B (S. cerevisiae); SEC23-like protein B; SEC23-related protein B; transport protein SEC23B | |
Mouse | |
Affinity Chromatography | |
RUO | |
10483 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction