Learn More
SEC24B Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304876AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
Component of the coat protein complex II (COPII) which promotes the formation of transport vesicles from the endoplasmic reticulum (ER). The coat has two main functions, the physical deformation of the endoplasmic reticulum membrane into vesicles and the selection of cargo molecules for their transport to the Golgi complex. Plays a central role in cargo selection within the COPII complex and together with SEC24A may have a different specificity compared to SEC24C and SEC24D. May package preferentially cargos with cytoplasmic DxE or LxxLE motifs and may also recognize conformational epitopes.Specifications
SEC24B | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
O95487 | |
Sec24b | |
Between 1 and 50 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
SEC24B | |
SEC24, SEC24 (S. cerevisiae) related gene family, member B, SEC24 family member B, SEC24 family, member B (S. cerevisiae), SEC24 related gene family, member B, SEC24-related protein B, secretory protein 24 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10427, 99683 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.