Learn More
SEC24C Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 160731AP
Catalog No. 50-173-0692
Add to cart
Description
Description
Interacts with HSP70 and can stimulate its ATPase activity. Stimulates the association between HSC70 and HIP.
Specifications
Specifications
SEC24C
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P53992
KIAA0079, SEC24 related protein C, SEC24C
Rabbit
IgG
150 μL
-20°C
Primary
218811, 685144, 9632
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
SEC24C
Liquid
Sec24c
SEC24C Fusion Protein Ag9030
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat