Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SEC31A Rabbit anti-Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP17957520UL

 View more versions of this product

Catalog No. NBP17957520


Add to cart

Description

Description

SEC31A Polyclonal antibody specifically detects Antigen in Rat samples. It is validated for Western Blotting.
Specifications

Specifications

SEC31A
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Affinity Purified
SEC31A
Synthetic peptide directed towards the N terminal of human Sec31aThe immunogen for this antibody is Sec31a. Peptide sequence DKEVVIAQKDKHTGPVRALDVNIFQTNLVASGANESEIYIWDLNNFATPM.
136 kDa
20ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Rat
Western Blot
Western Blot 1:1000
NP_148981
ABP125ABP130KIAA0905Web1-like protein, DKFZp686N07171, HSPC334, MGC90305, protein transport protein Sec31A, protein-transport protein SEC31, SEC31 homolog A (S. cerevisiae), SEC31L1, SEC31-like 1, SEC31-like 1 (S. cerevisiae), SEC31-like protein 1, SEC31-related protein A, yeast Sec31p homolog
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
22872
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit