SecinH3, Tocris

Sec7-specific GEF inhibitor (cytohesins)

Manufacturer: R&D Systems 2849/50

Quantity 50mg

 View more versions of this product

Catalog No. 28-495-0

  Check Availability
Add to Cart

    Chemical Identifiers

    Assay Percent Range >99%
    Molecular Formula C24H20N4O4S
    Formula Weight 460.5
    Synonym N-[4-[5-(1,3-Benzodioxol-5-yl)-3-methoxy-1H-1,2,4-triazol-1-yl]phenyl]-2-(phenylthio)acetamide
    Chemical Name or Material SecinH3

    Specifications

    Formula Weight 460.5
    Quantity 50mg
    Synonym N-[4-[5-(1,3-Benzodioxol-5-yl)-3-methoxy-1H-1,2,4-triazol-1-yl]phenyl]-2-(phenylthio)acetamide
    Assay Percent Range >99%
    Molecular Formula C24H20N4O4S
    CAS 853625-60-2
    Recommended Storage Store at +4°C
    Chemical Name or Material SecinH3

    Store at +4°C

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit