Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SecinH3, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Sec7-specific GEF inhibitor that displays selectivity for small GEFs of the cytohesin family (IC50 values are 2.4, 5.4, 5.4, 5.6, 5.6, 65 and > 100 μM for hCyh2, hCyh1, mCyh3, hCyh3, drosophila steppke, yGea2-S7 and hEFA6-S7 respectively). Inhibits insulin signaling via inhibition of insulin receptor substrate protein (IRS) phosphorylation.
Specifications
Quantity | 10 mg |
Formula Weight | 460.5 |
Percent Purity | >99% |
Chemical Name or Material | ARF GTPase Inhibitors |
Safety and Handling
Recommended Storage : Store at +4°C