Learn More
SECO™ Rhino Shield™ 510 Foil Bags
Supplier:  SECO™ 51068
Encompass
 View more versions of this product
Catalog No. 19-045-628
Add to Cart
Description
Description
The combination of polyethylene for heat sealing and foil for vacuum retention produces a laminated-film material with superior barrier properties against water vapor and oxygen. Flexible, with excellent tensile strength, and protect against atmospheric changes as well as water, grease, acid, chemicals, and corrosion.
Order Info
Available in three sizes. Packaged 100 per pack.
Specifications
Specifications
Laminated Polyethylene, Foil
8 in.
6 in.
8 x 6 in. (20 x 15cm)
20 cm
15 cm
Product Suggestions
Product Suggestions