Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Secretagogin Antibody Pair [HRP]

Mouse Monoclonal Antibody Pair

Manufacturer:  Novus Biologicals™ NBP2793965PLATES

 View more versions of this product

Catalog No. NB009832



Description

Description

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Secretagogin.
Specifications

Specifications

Sandwich ELISA
Antibody Pairs
7.8 pg/mL
10590
RUO
Secretagogin
5 Plates
7.81-500 pg/mL
CALBL, DJ501N12.8, secretagogin, EF-hand calcium binding protein, SECRETsecretagogin, SEGN, setagin
HRP
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Secretagogin.
Sandwich ELISA
  • Standard: Each vial contains 21 ng of recombinant Secretagogin.
  • Capture Antibody: 0.1 mg/mL of mouse anti-Secretagogin monoclonal antibody.
  • Detection Antibody: 0.5 mg/mL mouse anti-Secretagogin monoclonal antibody conjugated to horseradish-peroxidase (HRP).
  • Storage is content dependent.
    Mouse
    Human
    SCGN
    1. Capture Antibody: Dilute to a working concentration of 2.0 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 1 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute standard powder with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 500 pg/mL is recommended.
    SDS
    Documents
    Product Certifications
    Promotions

    Promotions

    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit