Learn More
Secretagogin Rabbit anti-Human, Alexa Fluor 350, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP299985AF350
Catalog No. NB063871
Add to cart
Description
Description
Secretagogin Polyclonal antibody specifically detects Secretagogin in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
Specifications
Secretagogin
Alexa Fluor 350
50mM Sodium Borate
CALBL, DJ501N12.8, secretagogin, EF-hand calcium binding protein, SECRETsecretagogin, SEGN, setagin
Produced in rabbits immunized with purified, recombinant Secretagogin (NP_008929.2; Asp 2-Pro 276).
Affinity purified
RUO
Primary
10590
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
Rabbit
IgG
0.1 mL
Store at 4°C in the dark.
Polyclonal
Human