Learn More
Secretagogin Rabbit anti-Human, Biotin, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP299985B
Catalog No. NB063879
Add to cart
Description
Description
Secretagogin Polyclonal specifically detects Secretagogin in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
Secretagogin
Biotin
PBS
CALBL, DJ501N12.8, secretagogin, EF-hand calcium binding protein, SECRETsecretagogin, SEGN, setagin
Produced in rabbits immunized with purified, recombinant Secretagogin (NP_008929.2; Asp 2-Pro 276).
Affinity purified
RUO
Primary
10590
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
Rabbit
IgG
0.1 mL
Store at 4°C in the dark.
Polyclonal
Human