Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Secretory Component Ab-2 Mouse Monoclonal Antibody, 1.9mg/mL, BSA and azide

Provide accurate, reproducible results with the Epredia™ Secretory Component Ab-2, Mouse Monoclonal Antibody.

Supplier:  Epredia™ MS1367R1

 View more versions of this product

Catalog No. MS1367R1

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.


Description

Description

Secretory Component is an epithelial transport receptor for polymeric IgA and IgM. Antibody to secretory component has been shown to be useful for identifying glandular carcinomas.

Host Species: Mouse

Clone: COMPO2; same as SC-05

Isotype: IgG1

Species Reactivity: Human and Rat. Others not known.

Epitope: Not determined

Immunogen: Semi-purified human secretory component (SC)

Molecular Weight: 80kDa

Positive Control: Small intestine, Colon CA, Stomach

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Secretory Component Ab-2
Monoclonal
Unconjugated
Semi- purified human secretory component (SC)
RUO
Primary
Prediluted
Immunofluorescence, Immunohistochemistry (Paraffin)
COMPO2; same as SC-05
Mouse
1 mL
Cancer and Tumor Biology
Human, Rat
IgG1
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit