Brady™ SecurAlert Clip-On Badges

Prevent unauthorized use of visitor, contractor and temporary passes. Brady™ SecurAlert Clip-On Badges are self-expiring, which prevents unauthorized use beyond their intended use periods.

Manufacturer: Brady™ 95668

 View more versions of this product

Catalog No. 19-103-170

  Check Availability
Add to Cart

    Specifications

    Color Red
    Dimensions (L x W) 3 in. x 3 in.
    For Use With (Application) Temporary
    Item Description 1 week expiration
    Width (English) 3 in.
    Length (English) 3 in.

    • Self-expiring badges
    • Low cost clip-on alternative to self-adhesive badges
    • 3 in. x 3 in. slotted cards
    • 500 front and back badge pairs per pack

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit