Learn More
AVANTOR MASTERFLEX (VWR LLC) 155751-43 Double open end polypropylene sediment cartridge; 25 m, 10 inch length
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX0151228
Encompass_Preferred
Ideal for removing a broad range of deformable and nondeformable particles. Rigid design is ideal for filtering high-solids, higher-viscosity fluids that require higher differential pressures. Cartridge imparts no taste, odor, or color. Maximum temperature is 175¿F (79¿C).
Note: For optimal performance, change cartridges at 25 psid.
Catalog No. 50-632-099