Learn More
Tocris Bioscience™ Seglitide
sst2 and sst5 agonist
Manufacturer:  Tocris Bioscience™ 1836/1
Catalog No. 18-361-R
Add to cart
This item is not returnable. View return policy
Description
Description
Peptide agonist for sst2 and sst5 somatostatin receptors. IC50/Kd values (nM) at cloned human somatostatin receptors are: > 1000 (sst1), 0.2 - 1.5 (sst2), 27 - 36 (sst3), > 127 (sst4), and 0.06 - 23 (sst5).
Specifications
Specifications
Seglitide
1mg
Seglitide
808.98
cyclo(N-methyl-Ala-Tyr-D-Trp-Lys-Val-Phe)