Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Segmentation protein paired Rabbit anti-Drosophila, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$482.50
Specifications
Antigen | Segmentation protein paired |
---|---|
Dilution | Western Blot 1.0 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB123411
|
Novus Biologicals
NBP309255100UL |
100 μg |
Each of 1 for $482.50
|
|
|||||
Description
Segmentation protein paired Polyclonal specifically detects Segmentation protein paired in Drosophila samples. It is validated for Western Blot.Specifications
Segmentation protein paired | |
Western Blot | |
Unconjugated | |
Rabbit | |
Drosophila | |
PRD | |
The immunogen is a synthetic peptide corresponding to a region of Fruit fly (NP_723721). Peptide sequence MTVTAFAAAMHRPFFNGYSTMQDMNSGQGRVNQLGGVFINGRPLPNNIRL | |
Affinity purified |
Western Blot 1.0 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
PBS buffer, 2% sucrose | |
34629 | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title