Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Black Stallion™ Select Shoulder Split Cowhide Leather Welder's Gloves with Cotton Lining

Split cowhide leather gloves feature cotton sock lining for comfort, and Aramid Fiber stitching for spark and abrasion resistance.

$27.50 - $338.50

Specifications

Color Orange
For Use With (Application) Stick Welding
Glove Cuff Style Safety
Liner Material Cotton
Material Shoulder Split Cowhide Leather
View More Specs

Products 2
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-063-117
View Documents
Black Stallion™
110S
Small
Pair for $27.50
Add to cart
 
18-999-1312
View Documents
Black Stallion™
110L
Large
Case of 12 for $338.50
Add to cart
 
Description

Description

  • Select shoulder split cowhide leather construction is reliably consistent
  • Cotton sock lining for additional comfort and thermal protection
  • Aramid Fiber stitching for spark and abrasion resistance
  • Thumb strap reinforcement provides extra durability
  • Ideal protection for standard stick welding applications
Specifications

Specifications

Orange
Safety
Shoulder Split Cowhide Leather
Stick Welding
Cotton
110
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit