Selecto™ Flexible-Backed TLC Plates
Select from four media to match your separation needs
Manufacturer: Selecto ScientificSupplier Diversity Partner 30028
 View more versions of this product
Catalog No. 05-719-858
  Check Availability
Add to Cart
    Specifications
    Product Type Silica TLC plate
    Backing Material Plastic
    For Use With (Application) Widely used for Dyes, Heterocyclic's, Pesticides, Herbicides, Fatty Acids, Amino Acids, various Alcohols, Steroids, Hydrocarbons and Organic synthesis compounds
    No. per Pack 50.0
    Quantity 50/Pack
    Size (English) 1.96 x 7.87 in.
    Size (Metric) 5 x 20cm
    Thickness (Metric) 200μm
    Sorbent Silica gel
  • Description & Specifications
Related Products
Product Content Feedback