Selecto™ Flexible-Backed TLC Plates
Select from four media to match your separation needs
Manufacturer: Selecto ScientificSupplier Diversity Partner 11045
 View more versions of this product
Catalog No. 05-719-862
  Check Availability
Add to Cart
    Specifications
    Product Type Alumina TLC plate
    Backing Material Plastic
    For Use With (Application) Widely used for Alkaloids, Steroids, Terpenes, aliphatic and aromatic compounds
    Length (Metric) 20cm
    No. per Pack 25.0
    Width (Metric) 20cm
    Quantity 25/Pack
    Thickness (Metric) 200μm
    Sorbent Alumina Basic
  • Description & Specifications
Related Products
Product Content Feedback