Selecto™ Flexible-Backed TLC Plates
Select from four media to match your separation needs
Manufacturer: Selecto ScientificSupplier Diversity Partner 10028
 View more versions of this product
Catalog No. 05-719-856
  Check Availability
Add to Cart
    Specifications
    Product Type Silica TLC plate
    Backing Material Plastic
    For Use With (Application) Widely used for Dyes, Heterocyclic's, Pesticides, Herbicides, Fatty Acids, Amino Acids, various Alcohols, Steroids, Hydrocarbons and Organic synthesis compounds
    Length (Metric) 20cm
    No. per Pack 25.0
    Quantity 25/Pack
    Thickness (Metric) 200μm
    Width (Metric) 20cm
    Sorbent Silica gel
  • Description & Specifications
Related Products
Product Content Feedback