Selecto™ Preparative and Classic Column Chromatography Active Silica
60Å gels made from identical starting material, making scale-up easier
Manufacturer: Selecto ScientificSupplier Diversity Partner 142824
 View more versions of this product
Catalog No. 05-719-824
  Check Availability
Add to Cart
    Specifications
    pH 7
    Quantity 500g
    Material Silica Gel
    Particle Size 32 to 63μm
  • Description & Specifications
Related Products
Product Content Feedback