Learn More
Aviva Systems Biology Corp SELENBP1 Antibody - N-terminal region (ARP48233_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP48233_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SELENBP1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SELENBP1. Alternate names: FLJ13813, LPSB, SP56, hSBP, hSP56, SBP56ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59770