Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SELENBP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA537332
Description
This antibody detects endogenous protein at a molecular weight of 44 and 53 kDa. Purity is >95% by SDS-PAGE.
Selenium is an essential trace element that confers tolerance to toxicity arising through exposure to heavy metals or other reactive xenobiotics. Selenium exhibits potent anticarcinogenic properties, and deficiency of selenium may cause certain neurologic diseases. Both effects are attributed to selenium-binding proteins. Selenium binding protein 1 is down-regulated in lung adenocarcinoma, colorectal cander and ovarian cancer. It is two-fold upregulated in the brains of patients suffering from schizophrenia, and is therefore a biomarker for this disease.
Specifications
SELENBP1 | |
Polyclonal | |
Unconjugated | |
SELENBP1 | |
56 kDa selenium-binding protein; epididymis secretory sperm binding protein Li 134P; FLJ13813; HEL-S-134P; hSBP; hSP56; Lp 56; Lp56; LPSB; Methanethiol oxidase; methanethiol oxidase; selenium-binding protein 1; MTO; SBP; SBP 1; SBP1; SBP56; Selen bp2; SELENBP 1; Selenbp 2; selenbp1; Selenbp2; selenium binding protein 1; selenium binding protein 2; selenium-binding protein 1; Selenium-binding protein 2; SELNBP1; similar to selenium binding protein 1; SP 56; SP56; zgc:65844 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
20341, 8991 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P17563, Q13228 | |
SELENBP1 | |
Recombinant full length human SELENBP1. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction