Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Selenious acid, 98%, ACROS Organics™

Click to view available options
Quantity:
50g
250g
1kg
Packaging:
Glass bottle
7783-00-8
H2O3Se
128.97
MCAHWIHFGHIESP-UHFFFAOYSA-N
selenious acid,selenious acid h2seo3,monohydrated selenium dioxide,unii-f6a27p4q4r,selenium dioxide, monohydrated,ccris 5530,selenious acid usp,selenious acid,selenous acid,monohydrated selenium dioxide,selenium, reference standard solution
1091
CHEBI:26642
selenous acid
O[Se](=O)O
7783-00-8
H2O3Se
128.97
MCAHWIHFGHIESP-UHFFFAOYSA-N
selenious acid,selenious acid h2seo3,monohydrated selenium dioxide,unii-f6a27p4q4r,selenium dioxide, monohydrated,ccris 5530,selenious acid usp,selenious acid,selenous acid,monohydrated selenium dioxide,selenium, reference standard solution
1091
CHEBI:26642
selenous acid
O[Se](=O)O
Assay Percent Range 97.5% min. (Iodometry)
Fieser 01,992
Merck Index 15,8567
Solubility Information Solubility in water: 167g/100mL (20°C). Other solubilities: very soluble in hot water and alcohol,insoluble in ammonia
Formula Weight 128.97
Physical Form Crystalline Powder, Crystals and/or Chunks
Percent Purity 98%
Packaging Glass bottle
Solubility Solubility in water: 167 g/100mL (20°C). Other solubilities: very soluble in hot water and alcohol, insoluble in ammonia
Color Pink or White
Melting Point 70.0°C
Quantity 250g
Chemical Name or Material Selenious acid
Show More Show Less

GHS Signal Word: Danger

EINECSNumber : 231-974-7