Learn More
Thermo Scientific™ Selenite Broth
Description
Reduce media preparation time and simultaneously enrich and isolate Salmonella spp. using convenient, pre-filled, ready-to-use Thermo Scientific™ Selenite Broth.
Remove the need for pipetting, dispensing or autoclaving with convenient, pre-filled and ready-to-use tubes of Selenite Broth.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

Specifications
Specifications
Color | Clear liquid, faint to very light amber |
Content And Storage | 2°C to 8°C |
Format | Tube |
Manufacturing Origin | United States |
Media Type | Selenite Broth |
Product Type | Microbiology Media |
Fill Volume | 9 mL |
Packaging Type | Glass tube with black screw cap |
Quantity | 100/Pk. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.