Selenium, 1000µg/mL (0.10% w/v), BAKER INSTRA-ANALYZED™ Reagent, J.T.Baker™

Acids and standards that are purified and characterized for use in trace element analysis in the low parts-per-billion (ppb) range

Manufacturer: J.T.Baker 577704

Packaging Poly Bottle
Quantity 100mL

 View more versions of this product

Catalog No. 02-003-886

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    Molecular Formula Se
    Formula Weight 78.96
    Chemical Name or Material Selenium, 1000μg/mL (0.10% w/v)
    Grade BAKER INSTRA-ANALYZED™ Reagent

    Specifications

    Formula Weight 78.96
    Specific Gravity 1L = 1.00kg
    Quantity 100mL
    Molecular Formula Se
    Packaging Poly Bottle
    Chemical Name or Material Selenium, 1000μg/mL (0.10% w/v)
    Grade BAKER INSTRA-ANALYZED™ Reagent

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit