Learn More
Selenium(IV) oxide, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Description
In organic reactions, selenium dioxide is often used as an oxidizing reagent employed for carrying out allylic oxidations, Riley oxidation, synthesis of diones, etc. It is used to impart a red color to glass. It finds use as a toner in photographic developing, and an active ingredient in some cold-blueing solutions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 340°C to 350°C (sublimation) |
Physical Form | Lump/Powder |
Quantity | 5 g |
UN Number | UN3283 |
Merck Index | 14,8434 |
Solubility Information | Soluble in water,benzene,ethanol and methanol. Sprangly soluble in acetone and acetic acid. |
Formula Weight | 110.96 |
Percent Purity | 99.999% |
Grade | Puratronic™ |
Odor | Odorless |
Show More |
Safety and Handling
Hazard Category | H301-H310+H330-H314-H341-H373 |
Hazard Statement | GHS H Statement H301-H331-H373 Toxic if swallowed. Toxic if inhaled. May cause damage to organs through prolonged or repeated exposure. |
Precautionary Statement | P201-P202-P261-P262-P264b-P270-P280-P285-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P310-P330-P363-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: SELENIUM COMPOUND, SOLID, N.O.S. |
EINECSNumber | 231-194-7 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.