Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Selenium, Oil based standard solution, Specpure™ Se 1000μg/g

Catalog No. AA4530518
Click to view available options
Quantity:
50 g

Catalog No. AA4530518

Supplier: Thermo Scientific Chemicals 04530518

Only null left
Add to Cart

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00134090

Specifications

Physical Form Liquid
Quantity 50 g
Grade Specpure™
Chemical Name or Material Selenium, Oil based standard solution, Se 1000μg/g
Hazard Category Danger
Hazard Statement GHS H Statement
H301-H373-H332
Toxic if swallowed.
May cause damage to organs through prolonged or repeated exposure.
Harmful if inhaled.
Precautionary Statement GHS P Statement
P260-P261-P301+P310-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.