Selenium shot, 99.99% (metals basis), Alfa Aesar™, Puratronic™

Manufacturer: Alfa Aesar 1060318

Quantity 50g

 View more versions of this product

Catalog No. AA1060318

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7782-49-2
    Physical Form Shot, Amorphous, 2 To 6mm (0.08 To 0.2 in.)
    Assay Percent Range 99.999% (Metals basis)
    MDL Number MFCD00134090
    Chemical Name or Material Selenium

    Specifications

    Physical Form Shot, Amorphous, 2 To 6mm (0.08 To 0.2 in.)
    Quantity 50g
    Density 4.28g/cm3
    EINECS Number 231-957-4
    Assay Percent Range 99.999% (Metals basis)
    Appearance Shot
    MDL Number MFCD00134090
    CAS 7782-49-2
    Health Hazard 1 H301-H331-H373-H413: Toxic if swallowed. Toxic if inhaled. May cause damage to organs through prolonged or repeated exposure. May cause long lasting harmful effects to aquatic life.
    Chemical Name or Material Selenium

    H301-H331-H373-H413: Toxic if swallowed. Toxic if inhaled. May cause damage to organs through prolonged or repeated exposure. May cause long lasting harmful effects to aquatic life.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit