Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Selenium Shot, amorphous, 1 to 5mm (0.04 to 0.20in.), 99.999% (Metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 0029030

Quantity 250g

 View more versions of this product

Catalog No. AA0029030


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7782-49-2
>99.999% (Metals basis)
Selenium
Shot, Amorphous, Solid In Various Forms, 1 To 5mm (0.04 To 0.20 in.)
MFCD00134090

Specifications

7782-49-2
>99.999% (Metals basis)
MFCD00134090
Various
Selenium
Shot, Amorphous, Solid In Various Forms, 1 To 5mm (0.04 To 0.20 in.)
Various colors shot
Odorless
250g

Safety and Handling

SDS & Safety Documents

H301-H331-H373-H413: Toxic if swallowed. Toxic if inhaled. May cause damage to organs through prolonged or repeated exposure. May cause long lasting harmful effects to aquatic life.

EINECSNumber : 231-957-4

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit