Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Selenium Sulfide, USP, 52-55.5%, Spectrum™

Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
5 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
7488-56-4
S2Se
143.09
JNMWHTHYDQTDQZ-UHFFFAOYSA-N
thione
S=[Se]=S
7488-56-4
S2Se
143.09
JNMWHTHYDQTDQZ-UHFFFAOYSA-N
thione
S=[Se]=S

S1019, 7488-56-4, SeS2

Spectrum™ Selenium Sulfide, USP is used to treat dandruff and a certain scalp infections. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Grade USP
CAS Max % 1
Percent Purity 52 to 55.5%
Ignition Residue 0.002
Packaging Poly Bottle
Quantity 500 g