Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Selenophene 98.0+%, TCI America™
Click to view available options
Quantity:
1g
5g
Specifications
Color | Yellow |
Melting Point | -30°C |
Boiling Point | 110°C |
UN Number | 1992 |
Quantity | 5g |
Formula Weight | 131.04 |
Physical Form | Clear Liquid at 20°C |
Percent Purity | ≥98.0% (GC) |
Chemical Name or Material | Selenophene |
Safety and Handling
TSCA : No