Selenous acid, 97%, Alfa Aesar™

For Use As An Oxidizing Agent, As a Reagent For Alkaloids

Manufacturer: Alfa Aesar 1257430

Quantity 250g

 View more versions of this product

Catalog No. AA1257430

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7783-00-8
    Physical Form Crystalline
    Assay Percent Range 97%
    Molecular Formula H2SeO3
    Formula Weight 128.97
    MDL Number MFCD00011331
    Synonym Selenious acid
    Chemical Name or Material Selenous acid

    Specifications

    Physical Form Crystalline
    Formula Weight 128.97
    Quantity 250g
    Color Colorless
    DOT Information DOT Class: 6.1, Packaging Group: II
    EINECS Number 231-974-7
    Molecular Formula H2SeO3
    MDL Number MFCD00011331
    CAS 7783-00-8
    Health Hazard 1 H301-H331-H373-H400-H410: Toxic if swallowed. Toxic if inhaled. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.
    Odor Odorless
    Assay Percent Range 97%
    Synonym Selenious acid
    Chemical Name or Material Selenous acid

    H301-H331-H373-H400-H410: Toxic if swallowed. Toxic if inhaled. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit