Learn More
Brady™ Self-Adhesive Pipe Marker: VAPOR, Vinyl, Black on Yellow, Size 3C: Fits Pipes 0.75 in. Dia.
High gloss, durable, flexible vinyl with a permanent cold temperature pressure sensitive acrylic adhesive
Manufacturer:  Brady™ 83607
Catalog No. 19-112-107
Add to cart
Description
Description
  • High gloss, durable, flexible vinyl with a permanent cold temperature pressure sensitive acrylic adhesive
  • Good chemical, oil and water resistance
Specifications
Specifications
No
Black on Yellow
Pipe Marker
2.25 in. H x 2.75 in. W
2.25 in.
Card of 3 Each
0.010
350.52 mm
Pipe Marker
Pipe Marker
Permanent Acrylic
ANSI A13.1-2007
2.25 in. H x 2.75 in. W
0.5 in.
Card of 3 Each
2.25H x 2.75 in. W
2.75 in.
Spanish
Vinyl
Card of 3 Each