Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Self-Laminating, Two-Sided Polyester Tag: TARGET ORGANS AFFECTED: HEALTH HAZARD: FIRE HAZARD: INSTABILITY: CONTACT CORRESPONDING MSDS FOR FURTHER HAZARDOUS INFORMATION AND INSTRUCTIONS

Prevent your text from smearing or smudging in some of the harshest environments. Brady™ Self-Laminating, Two-Sided Polyester Tags are durable two-sided tags with a write-on area that accepts most marking pens.

Manufacturer:  Brady™ 52176

Catalog No. 19-113-091


Add to cart

Description

Description

  • Polyester tags provide identification solutions for almost any environment
  • Metal grommet prevents tearing from the point of attachment
  • Tag includes a write-on area that accepts most marking pens
  • Tags are appropriate for indoor or outdoor use
  • Tags are abrasion and chemical resistant
Specifications

Specifications

OSHA 1910.145
3 in. x 5.75 in.
Self-laminating
Rectangle
Polyester
Black/blue/red/yellow on white
(5) nylon fasteners
5/Pack
-40°F to 212°F
TARGET ORGANS AFFECTED: HEALTH HAZARD: FIRE HAZARD: INSTABILITY: CONTACT CORRESPONDING MSDS FOR FURTHER HAZARDOUS INFORMATION AND INSTRUCTIONS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit